missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline-2-carboxylic acid, 97%

Catalog No. AAH59780MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59780MC 100 mg
AAH59780ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59780MC Supplier Thermo Scientific Chemicals Supplier No. H59780MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 879-65-2 | C9H6N2O2 | 174.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 879-65-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.16
MDL Number MFCD00012334
InChI Key UPUZGXILYFKSGE-UHFFFAOYSA-N
Synonym 2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
PubChem CID 96695
IUPAC Name quinoxaline-2-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC=CC2=N1

Specifications

Melting Point 208°C (decomposition)
Quantity 100 mg
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Quinoxaline-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only