missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinoline Yellow, 95%, pure, water soluble

Catalog No. AC277340250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC277340250 25 g Glass bottle
AC277341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC277340250 Supplier Thermo Scientific Chemicals Supplier No. 277340250
Only null left

CAS: 8004-92-0 | C18H9NNa2O8S2 | 477.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 8004-92-0
Molecular Formula C18H9NNa2O8S2
Molecular Weight (g/mol) 477.37
MDL Number MFCD00080727
InChI Key FZUOVNMHEAPVBW-UHFFFAOYSA-L
PubChem CID 129893443
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Specifications

Assay Percent Range 95%
Solubility Information Solubility in water: 225g/L (20°C).
Infrared Spectrum Authentic
IUPAC Name disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
Percent Purity 95%
Grade Pure
Packaging Glass bottle
Quantity 25 g
Physical Form Powder
Chemical Name or Material Quinoline Yellow
Hazard Category Warning
Hazard Statement GHS P Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only