missing translation for 'onlineSavingsMsg'
Learn More

Quinoline 97.0+%, TCI America™

Catalog No. Q008525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q008525G 25 g
Q0085500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q008525G Supplier TCI America Supplier No. Q008525G

Chemical Identifiers

CAS 91-22-5
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
InChI Key SMWDFEZZVXVKRB-UHFFFAOYSA-N
Synonym chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
PubChem CID 7047
ChEBI CHEBI:17362
IUPAC Name quinoline
SMILES C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point -15°C
Boiling Point 238°C
Quantity 25 g
UN Number 2656
Formula Weight 129.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Quinoline
EINECSNumber (5)-0794
RTECSNumber VA9275000
TSCA Yes