missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline 98.0+%, TCI America™
$67.11 - $253.51
Chemical Identifiers
| CAS | 91-22-5 |
|---|---|
| Molecular Formula | C9H7N |
| Molecular Weight (g/mol) | 129.16 |
| MDL Number | MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 |
| InChI Key | SMWDFEZZVXVKRB-UHFFFAOYSA-N |
| Synonym | chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol |
| PubChem CID | 7047 |
| ChEBI | CHEBI:17362 |
| IUPAC Name | quinoline |
| SMILES | C1=CC=C2N=CC=CC2=C1 |
Chemical Identifiers
| 91-22-5 | |
| 129.16 | |
| SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
| 7047 | |
| quinoline |
| C9H7N | |
| MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
| chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
| CHEBI:17362 | |
| C1=CC=C2N=CC=CC2=C1 |
Specifications
| 91-22-5 | |
| Yellow | |
| C9H7N | |
| 25 g | |
| chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
| C1=CC=C2N=CC=CC2=C1 | |
| 129.16 | |
| CHEBI:17362 | |
| ≥98.0% (GC,T) | |
| Quinoline |
| -15°C | |
| 238°C | |
| MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
| 2656 | |
| SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
| quinoline | |
| 7047 | |
| 129.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-0794
RTECSNumber : VA9275000
TSCA : Yes