missing translation for 'onlineSavingsMsg'
Learn More

Quinoline, 99%

CAS: 91-22-5 | C9H7N | 129.16 g/mol

Supplier:  Thermo Scientific Chemicals 221141000

 View more versions of this product

Catalog No. AC221141000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Quinoline
91-22-5
Gray to Purple
237.0°C
Authentic
Glass bottle
1.6230 to 1.6260
100 g
01,975; 02,356; 09,403
15,8185
Solubility in water: slightly soluble. Other solubilities: soluble in hot water,miscible with alcohol,ether,acetone,benzene,and carbon disulfide
C1=CC=C2N=CC=CC2=C1
129.16
CHEBI:17362
99%
99%
-15.0°C
1.0950g/mL
101°C
98.5% min. (GC)
C9H7N
MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
20,339
1.095
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
SMWDFEZZVXVKRB-UHFFFAOYSA-N
quinoline
7047
129.16
Liquid

Chemical Identifiers

91-22-5
129.16
SMWDFEZZVXVKRB-UHFFFAOYSA-N
7047
quinoline
C9H7N
MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
CHEBI:17362
C1=CC=C2N=CC=CC2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful in contact with skin.
Toxic

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENT

GHS Signal Word: Danger

EINECSNumber : 202-051-6

RTECSNumber : VA9275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only