Learn More
Quinoline, 99%
CAS: 91-22-5 | C9H7N | 129.16 g/mol
Supplier: Thermo Scientific Chemicals 221140025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Quinoline | |
| 91-22-5 | |
| Gray to Purple | |
| 237.0°C | |
| Authentic | |
| Glass bottle | |
| 1.6230 to 1.6260 | |
| 2.5 kg | |
| 01,975; 02,356; 09,403 | |
| 15,8185 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in hot water,miscible with alcohol,ether,acetone,benzene,and carbon disulfide | |
| C1=CC=C2N=CC=CC2=C1 | |
| 129.16 | |
| CHEBI:17362 | |
| 99% |
| 99% | |
| -15.0°C | |
| 1.0950g/mL | |
| 101°C | |
| 98.5% min. (GC) | |
| C9H7N | |
| MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
| 20,339 | |
| 1.095 | |
| chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
| SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
| quinoline | |
| 7047 | |
| 129.16 | |
| Liquid |
Chemical Identifiers
| 91-22-5 | |
| 129.16 | |
| SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
| 7047 | |
| quinoline |
| C9H7N | |
| MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
| chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
| CHEBI:17362 | |
| C1=CC=C2N=CC=CC2=C1 |
Safety and Handling
GHS H Statement
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful in contact with skin.
Toxic
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENT
GHS Signal Word: Danger
EINECSNumber : 202-051-6
RTECSNumber : VA9275000
TSCA : TSCA
RUO – Research Use Only