missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-sulfonic acid, 98%

Catalog No. AAL0664018
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0664018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0664018 Supplier Thermo Scientific Chemicals Supplier No. L0664018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85-48-3 | C9H7NO3S | 209.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-48-3
Molecular Formula C9H7NO3S
Molecular Weight (g/mol) 209.219
MDL Number MFCD00024037
InChI Key SXVRECLPTCOMIA-UHFFFAOYSA-N
Synonym 8-quinolinesulfonic acid, quinoline-8-sulphonic acid, unii-84y9b81o6x, ccris 6936, 8-sulfoquinoline, cambridge id 5135395, quinoline-8-sulfonic??acid, ksc242e8p, 8-quinolinesulfonic acid;quinoline-8-sulfonic acid
PubChem CID 66561
IUPAC Name quinoline-8-sulfonic acid
SMILES C1=CC2=C(C(=C1)S(=O)(=O)O)N=CC=C2

Specifications

Melting Point >300°C
Quantity 50 g
UN Number UN2585
Beilstein 9501
Formula Weight 209.23
Percent Purity 98%
Chemical Name or Material Quinoline-8-sulfonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 201-611-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only