missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-carboxaldehyde, 97%

Catalog No. aal1964206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1964206 5 g
AAL1964203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1964206 Supplier Thermo Scientific Chemicals Supplier No. L1964206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38707-70-9 | C10H7NO | 157.172 g/mol

Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 38707-70-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00805837
InChI Key OVZQVGZERAFSPI-UHFFFAOYSA-N
Synonym 8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
PubChem CID 170103
IUPAC Name quinoline-8-carbaldehyde
SMILES C1=CC2=C(C(=C1)C=O)N=CC=C2

Specifications

Melting Point 92°C to 94°C
Quantity 5 g
Beilstein 113039
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 157.17
Percent Purity ≥98%
Chemical Name or Material Quinoline-8-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only