missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7135645
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0086200MG 200 mg
Q00861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0086200MG Supplier TCI America Supplier No. Q0086200MG
Only null left

Chemical Identifiers

CAS 86-58-8
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.978
MDL Number MFCD01114698
InChI Key KXJJSKYICDAICD-UHFFFAOYSA-N
Synonym quinoline-8-boronic acid, 8-quinolineboronic acid, 8-quinolinylboronic acid, 8-quinoline boronic acid, quinolin-8-yl boronic acid, 8-boronoquinoline, 8-quinolylboronic acid, boronic acid, 8-quinolinyl, unii-5qs1a25ij6, quinolin-8-yl-8-boronic acid
PubChem CID 2734380
IUPAC Name quinolin-8-ylboronic acid
SMILES B(C1=C2C(=CC=C1)C=CC=N2)(O)O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 172.98
Physical Form Crystalline Powder
Chemical Name or Material Quinoline-8-boronic Acid (contains varying amounts of Anhydride)