Learn More
Quinoline-4-carboxaldehyde, 97+%
CAS: 4363-93-3 | C10H7NO | 157.17 g/mol
$165.22 - $699.05
Chemical Identifiers
| CAS | 4363-93-3 |
|---|---|
| Molecular Formula | C10H7NO |
| Molecular Weight (g/mol) | 157.17 |
| MDL Number | MFCD00006781 |
| InChI Key | MGCGJBXTNWUHQE-UHFFFAOYSA-N |
| Synonym | 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde |
| PubChem CID | 78072 |
| IUPAC Name | quinoline-4-carbaldehyde |
| SMILES | O=CC1=C2C=CC=CC2=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL2020306
|
Thermo Scientific Chemicals
L2020306 |
5 g |
Each for $165.22
|
|
|||||
|
AAL2020314
|
Thermo Scientific Chemicals
L2020314 |
25 g |
Each for $699.05
|
|
|||||
Description
4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store under dry inert gas. Incompatible with oxidizing agents and air.
Chemical Identifiers
| 4363-93-3 | |
| 157.17 | |
| MGCGJBXTNWUHQE-UHFFFAOYSA-N | |
| 78072 | |
| O=CC1=C2C=CC=CC2=NC=C1 |
| C10H7NO | |
| MFCD00006781 | |
| 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde | |
| quinoline-4-carbaldehyde |
Specifications
| 4363-93-3 | |
| 131°C to 133°C (5 mmHg) | |
| C10H7NO | |
| 5 g | |
| Air Sensitive | |
| Slightly soluble in water. | |
| O=CC1=C2C=CC=CC2=NC=C1 | |
| 157.17 | |
| 157.17 | |
| Quinoline-4-carboxaldehyde |
| 48°C to 52°C | |
| >110°C (230°F) | |
| MFCD00006781 | |
| 113072 | |
| 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde | |
| MGCGJBXTNWUHQE-UHFFFAOYSA-N | |
| quinoline-4-carbaldehyde | |
| 78072 | |
| ≥97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-453-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only