missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxaldehyde, 97+%

Catalog No. AAL2020314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2020314 25 g
AAL2020306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2020314 Supplier Thermo Scientific Chemicals Supplier No. L2020314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4363-93-3 | C10H7NO | 157.17 g/mol

4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store under dry inert gas. Incompatible with oxidizing agents and air.

Chemical Identifiers

CAS 4363-93-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00006781
InChI Key MGCGJBXTNWUHQE-UHFFFAOYSA-N
Synonym 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
PubChem CID 78072
IUPAC Name quinoline-4-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 48°C to 52°C
Boiling Point 131°C to 133°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 113072
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 157.17
Percent Purity ≥97%
Chemical Name or Material Quinoline-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-453-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only