missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-2-carbonitrile, 97%

Catalog No. AAL1942706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1942706 5 g
AAL1942703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1942706 Supplier Thermo Scientific Chemicals Supplier No. L1942706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1436-43-7 | C10H6N2 | 154.17 g/mol

Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1436-43-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00134341
InChI Key WDXARTMCIRVMAE-UHFFFAOYSA-N
Synonym 2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0
PubChem CID 74031
IUPAC Name quinoline-2-carbonitrile
SMILES N#CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 93°C to 96°C
Boiling Point 160°C (23 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 115213
Solubility Information Insoluble in water.
Formula Weight 154.17
Percent Purity 97%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material Quinoline-2-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement

H302-H315-H319-H335

Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-865-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only