missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline-2-carbonitrile, 97%
Description
Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 93°C to 96°C |
| Boiling Point | 160°C (23 mmHg) |
| Odor | Odorless |
| Quantity | 1 g |
| Beilstein | 115213 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 154.17 |
| Percent Purity | 97% |
| Physical Form | Crystals or Powder or Crystalline Powder |
| Chemical Name or Material | Quinoline-2-carbonitrile |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H StatementH302-H315-H319-H335 Harmful if swallowed.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 215-865-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only