missing translation for 'onlineSavingsMsg'
Learn More

Quinizarin 97.0+%, TCI America™

Catalog No. D024325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D024325G 25 g
D0243100G 100 g
D0243500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D024325G Supplier TCI America Supplier No. D024325G

Chemical Identifiers

CAS 81-64-1
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001209
InChI Key GUEIZVNYDFNHJU-UHFFFAOYSA-N
Synonym quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy
PubChem CID 6688
ChEBI CHEBI:37487
IUPAC Name 1,4-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O

Specifications

Melting Point 196°C
Boiling Point 450°C
Quantity 25 g
Formula Weight 240.21
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Quinizarin
EINECSNumber (4)-0704&(5)-5055
RTECSNumber CB6600000
TSCA Yes