missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinizarin Blue 90.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 329.36 |
| Percent Purity | ≥90.0% (HPLC) |
| Chemical Name or Material | Quinizarin Blue |
Safety and Handling
| EINECSNumber | (4)-0743&(5)-2587&(5)-3110 |
| RTECSNumber | CB7700000 |
| TSCA | Yes |