missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Quinine | |
| 177°C | |
| C20H24N2O2 | |
| 100 g | |
| LOUPRKONTZGTKE-ZCXAXGJJNA-N | |
| (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol | |
| 129316724 | |
| ≥98.0% (T) |
| 130-95-0 | |
| White | |
| MFCD00198096 | |
| 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol | |
| [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C | |
| 324.42 | |
| 324.42 | |
| Crystalline Powder |
Chemical Identifiers
| 130-95-0 | |
| 324.42 | |
| LOUPRKONTZGTKE-ZCXAXGJJNA-N | |
| 129316724 | |
| [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C |
| C20H24N2O2 | |
| MFCD00198096 | |
| 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol | |
| (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol |
Safety and Handling
EINECSNumber : (9)-0322
RTECSNumber : VA6020000
TSCA : Yes