Learn More
Thermo Scientific Chemicals Quinine sulfate dihydrate, 99+%
CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol
Supplier: Thermo Scientific Chemicals 163710100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Quinine sulfate dihydrate | |
| 6119-70-6 | |
| Beige to Brown or Yellow | |
| 3% to 5% (105°C) | |
| 99+% | |
| C40H54N4O10S | |
| 10 g | |
| −245.00 (20.00°C c=2,0.1M HCl) | |
| Solubility in water: 1g/810mL (20°C). Other solubilities: 1g/120mL alcohol,slightly soluble in chloroform and ether | |
| O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C | |
| 782.95 | |
| 782.96 | |
| Crystalline Powder |
| 99+% | |
| 233°C to 235°C | |
| 0.1 % max. | |
| Authentic | |
| Glass bottle | |
| MFCD00150790 | |
| 15, 8177 | |
| -245 | |
| ZHNFLHYOFXQIOW-OIGVVMIYNA-N | |
| (R)-[(2R,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;tetrahydrate | |
| 134129495 | |
| 99+% |
Chemical Identifiers
| 6119-70-6 | |
| 782.95 | |
| ZHNFLHYOFXQIOW-OIGVVMIYNA-N | |
| (R)-[(2R,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;tetrahydrate |
| C40H54N4O10S | |
| MFCD00150790 | |
| 134129495 | |
| O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
GHS Signal Word: Warning
RUO – Research Use Only