missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinine sulfate dihydrate, 99+%

Catalog No. AC163710500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163710500 50 g Glass bottle
AC163710100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163710500 Supplier Thermo Scientific Chemicals Supplier No. 163710500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6119-70-6
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00150790
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
PubChem CID 134129495
IUPAC Name (R)-[(2R,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;tetrahydrate
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point 233°C to 235°C
Color Beige to Brown or Yellow
Ignition Residue 0.1 % max.
Loss on Drying 3% to 5% (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 g
Merck Index 15, 8177
Specific Rotation Condition −245.00 (20.00°C c=2,0.1M HCl)
Specific Rotation -245
Solubility Information Solubility in water: 1g/810mL (20°C). Other solubilities: 1g/120mL alcohol,slightly soluble in chloroform and ether
Formula Weight 782.96
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Quinine sulfate dihydrate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.

RUO – Research Use Only