missing translation for 'onlineSavingsMsg'
Learn More

Quinine Sulfate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022950
Change view
Click to view available options
Quantity:
300 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022950 300 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022950 Supplier Merck Emd Millipore Supplier No. BP862

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 6119-70-6
Molecular Formula C20H24N2O2 · 0.5H2O4S · 2H2O
Linear Formula C20H24N2O2 · 0.5H2O4S · 2H2O
Quantity 300 mg
Synonym (8 alpha,9 R)- 6'-Methoxycinchonan-9-ol sulfate, hydrate (2:1:4)
Formula Weight 409.49
Grade BP