missing translation for 'onlineSavingsMsg'
Learn More

Quinine hemisulfate monohydrate, 98+%

Catalog No. AAA1703630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1703630 250 g
AAA1703609 10 g
AAA1703618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1703630 Supplier Thermo Scientific Chemicals Supplier No. A1703630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol

Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

Solubility
Soluble in a mixture of chloroform and absolute alcohol (2:1).

Notes
Light sensitive. Incompatible with strong oxidizing agents, alkalis, ammonia, strong bases and iodine.

Chemical Identifiers

CAS 6119-70-6
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00150790
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
Synonym Quinine sulfate (2:1) (salt) dihydrate; Quinine sulfate dihydrate
PubChem CID 134129495
IUPAC Name (R)-[(2R,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;tetrahydrate
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point 217°C to 218°C (decomposition)
Odor Odorless
Quantity 250 g
Beilstein 6113937
Sensitivity Light sensitive
Merck Index 14,8061
Solubility Information Soluble in a mixture of chloroform and absolute alcohol (2:1).
Optical Rotation −241° (c=2 in 0.1M HCl)
Formula Weight 391.47 (373.46 Anhydrous)
Percent Purity 99%
Chemical Name or Material Quinine hemisulfate monohydrate
Show More Show Less
Hazard Category H302+H312+H332-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H301-H334-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 212-359-2
RTECSNumber VA8440000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only