missing translation for 'onlineSavingsMsg'
Learn More

Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine

Catalog No. AAA1045909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1045909 10 g
AAA1045918 50 g
AAA1045930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1045909 Supplier Thermo Scientific Chemicals Supplier No. A1045909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 130-95-0 | C20H24N2O2 | 324.42 g/mol

Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.

Solubility
Soluble in alcohol, chloroform and diethyl ether. Slightly soluble in water and glycerol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 130-95-0
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00198096
InChI Key LOUPRKONTZGTKE-ZCXAXGJJNA-N
Synonym 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol
PubChem CID 129316724
SMILES [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point ∼177°C (decomposition)
Quantity 10 g
Beilstein 91867
Sensitivity Light Sensitive
Merck Index 14,8061
Solubility Information Soluble in alcohol,chloroform and diethyl ether. Slightly soluble in water and glycerol.
Optical Rotation −166° (c=1 in ethanol)
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Formula Weight 324.42
Percent Purity 99%
Assay (total base)
Chemical Name or Material Quinine, Anhydrous, may cont. up to 5% dihydroquinine
Show More Show Less
Hazard Category H302-H317
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 205-003-2
RTECSNumber VA6020000
TSCA Yes

RUO – Research Use Only