Learn More
(+)-Quinidine
CAS: 56-54-2 | C20H24N2O2 | 324.42 g/mol
Supplier: Thermo Scientific Chemicals A1255914
Description
(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
| (+)-Quinidine | |
| 171°C to 173°C | |
| MFCD00135581,MFCD00198096 | |
| UN2811 | |
| Light sensitive | |
| quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine | |
| LOUPRKONTZGTKE-LHHVKLHASA-N | |
| COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1 | |
| 324.42 | |
| CHEBI:28593 |
| 56-54-2 | |
| C20H24N2O2 | |
| 25 g | |
| 91866 | |
| 14,8060 | |
| Soluble in methanol,chloroform,alcohol (1g/36ml),benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water. | |
| +260° (c=1 in Ethanol) | |
| (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol | |
| 441074 | |
| 324.42 |
Chemical Identifiers
| 56-54-2 | |
| 324.42 | |
| LOUPRKONTZGTKE-LHHVKLHASA-N | |
| 441074 | |
| (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol |
| C20H24N2O2 | |
| MFCD00135581,MFCD00198096 | |
| quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine | |
| CHEBI:28593 | |
| COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
H301-H317
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-279-0
RTECSNumber : VA4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only