missing translation for 'onlineSavingsMsg'
Learn More

(+)-Quinidine

Catalog No. AAA1255903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1255903 1 g
AAA1255906 5 g
AAA1255914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1255903 Supplier Thermo Scientific Chemicals Supplier No. A1255903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56-54-2 | C20H24N2O2 | 324.42 g/mol

(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00135581,MFCD00198096
InChI Key LOUPRKONTZGTKE-LHHVKLHASA-N
Synonym quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
PubChem CID 441074
ChEBI CHEBI:28593
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1

Specifications

Melting Point 171°C to 173°C
Quantity 1 g
UN Number UN2811
Beilstein 91866
Sensitivity Light sensitive
Merck Index 14,8060
Solubility Information Soluble in methanol,chloroform,alcohol (1g/36ml),benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Formula Weight 324.42
Chemical Name or Material (+)-Quinidine
Hazard Category H301-H317
Hazard Statement GHS H Statement
H301
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-279-0
RTECSNumber VA4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only