missing translation for 'onlineSavingsMsg'
Learn More

Quinidine Sulfate Dihydrate 98.0+%, TCI America™

Catalog No. p-7135618
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00105G 5 g
Q001025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00105G Supplier TCI America Supplier No. Q00105G
Only null left

Chemical Identifiers

CAS 6591-63-5
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00135926
InChI Key ZHNFLHYOFXQIOW-CMZPEELOSA-N
Synonym bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate
PubChem CID 132274356
IUPAC Name (S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate
SMILES COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 746.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinidine Sulfate Dihydrate
EINECSNumber (9)-0324
RTECSNumber VA5605000
TSCA Yes