missing translation for 'onlineSavingsMsg'
Learn More

Quinidine sulfate dihydrate, 98+%

Catalog No. AAJ6042606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6042606 5 g
AAJ6042614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6042606 Supplier Thermo Scientific Chemicals Supplier No. J6042606

A sodium channel blocker, and to a lesser extent also blocks potassium channels | CAS: 6591-63-5 | C40H54N4O10S | 782.95 g/mol

Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.

Solubility
Soluble in chloroform, water (11 mg/ml at 20°C), alcohol, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away fom strong oxidizing agents.

Chemical Identifiers

CAS 6591-63-5
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00149346
InChI Key ZHNFLHYOFXQIOW-CMZPEELOSA-N
Synonym bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate
PubChem CID 132274356
IUPAC Name (S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate
SMILES COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O

Specifications

Melting Point 212°C to 214°C (decomposition)
Color White to Yellow
Odor Odorless
Quantity 5 g
Sensitivity Light sensitive
Merck Index 14,8060
Solubility Information Soluble in chloroform,water (11mg/ml at 20°C),alcohol,and ethanol (50mg/ml).
Formula Weight 782.94
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Quinidine sulfate dihydrate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-046-3
RTECSNumber VA5605000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only