missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quincorine 98.0+%, TCI America™
$765.02 - $765.02
Chemical Identifiers
| CAS | 207129-35-9 |
|---|---|
| Molecular Formula | C10H17NO |
| Molecular Weight (g/mol) | 167.252 |
| InChI Key | GAFZBOMPQVRGKU-FIBVVXLUSA-N |
| Synonym | (1S,2S,5S)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2S,5S)-5-Vinyl-2-quinuclidinemethanol |
| PubChem CID | 92020979 |
| IUPAC Name | [(2S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]methanol |
| SMILES | C=CC1CN2CCC1CC2CO |
Chemical Identifiers
| 207129-35-9 | |
| 167.252 | |
| (1S,2S,5S)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2S,5S)-5-Vinyl-2-quinuclidinemethanol | |
| [(2S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]methanol |
| C10H17NO | |
| GAFZBOMPQVRGKU-FIBVVXLUSA-N | |
| 92020979 | |
| C=CC1CN2CCC1CC2CO |
Specifications
| 207129-35-9 | |
| White-Yellow | |
| C10H17NO | |
| (1S,2S,5S)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2S,5S)-5-Vinyl-2-quinuclidinemethanol | |
| C=CC1CN2CCC1CC2CO | |
| 167.252 | |
| 167.25 | |
| Crystalline Lumps |
| 25°C | |
| 270°C | |
| 1 g | |
| GAFZBOMPQVRGKU-FIBVVXLUSA-N | |
| [(2S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]methanol | |
| 92020979 | |
| ≥98.0% (GC) | |
| Quincorine |
Safety and Handling
TSCA : No