missing translation for 'onlineSavingsMsg'
Learn More

Quinapril hydrochloride, 98%

Catalog No. p-7042265
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61913MC 100 mg
AAJ6191303 1 g
AAJ6191306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61913MC Supplier Thermo Scientific Chemicals Supplier No. J61913MC
Only null left

Angiotensin-converting enzyme inhibitor | CAS: 82586-55-8 | C25H31ClN2O5 | 474.982 g/mol

Angiotensin-converting enzyme inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82586-55-8
Molecular Formula C25H31ClN2O5
Molecular Weight (g/mol) 474.982
MDL Number MFCD00889215
InChI Key IBBLRJGOOANPTQ-JKVLGAQCSA-N
Synonym quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
PubChem CID 54891
ChEBI CHEBI:8714
IUPAC Name (3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
SMILES CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl

Specifications

Melting Point 120°C to 130°C
Color White
Quantity 100 mg
Merck Index 14,8051
Formula Weight 474.99
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Quinapril hydrochloride
Hazard Category H361
Precautionary Statement P201-P202-P281-P308+P313-P501c
RTECSNumber NW7176000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only