missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Quinapril hydrochloride, 97%

Catalog No. AAH3720903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3720903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3720903 Supplier Thermo Scientific™ Supplier No. H37209MD

Specifications

CAS 82586-55-8
Molecular Formula C25H31ClN2O5
MDL Number MFCD00889215
Quantity 1 g
InChI Key IBBLRJGOOANPTQ-JKVLGAQCSA-N
SMILES [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CC2=CC=CC=C2C[C@H]1C(O)=O
IUPAC Name hydrogen (3S)-2-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid chloride
Molecular Weight (g/mol) 474.98
Formula Weight 474.98
Percent Purity 97%
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 620-479-5
RTECSNumber NW7176000