missing translation for 'onlineSavingsMsg'
Learn More

Quinaldine Red 95.0+%, TCI America™

Catalog No. Q00071G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00071G 1 g
Q000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00071G Supplier TCI America Supplier No. Q00071G

Chemical Identifiers

CAS 117-92-0
Molecular Formula C21H23IN2
Molecular Weight (g/mol) 430.33
MDL Number MFCD00011968
InChI Key JOLANDVPGMEGLK-UHFFFAOYSA-M
PubChem CID 5702759
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
SMILES [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C

Specifications

Formula Weight 430.33
Percent Purity ≥95.0% (T)
Color Black/Green
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Quinaldine Red