missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinaldic Acid 98.0+%, TCI America™
Supplier: TCI America Q00035G
Specifications
| Quinaldic Acid | |
| 157°C | |
| C10H7NO2 | |
| 5 g | |
| LOAUVZALPPNFOQ-UHFFFAOYSA-N | |
| quinoline-2-carboxylic acid | |
| 7124 | |
| 173.17 | |
| Crystalline Powder |
| 93-10-7 | |
| White-Yellow | |
| MFCD00006752 | |
| quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid | |
| OC(=O)C1=CC=C2C=CC=CC2=N1 | |
| 173.17 | |
| CHEBI:18386 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 93-10-7 | |
| 173.17 | |
| LOAUVZALPPNFOQ-UHFFFAOYSA-N | |
| 7124 | |
| quinoline-2-carboxylic acid |
| C10H7NO2 | |
| MFCD00006752 | |
| quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid | |
| CHEBI:18386 | |
| OC(=O)C1=CC=C2C=CC=CC2=N1 |
Safety and Handling
EINECSNumber : (5)-0821
RTECSNumber : UZ9100000
TSCA : Yes