missing translation for 'onlineSavingsMsg'
Learn More

Quinaldic acid, 99%

Catalog No. AC171050100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171050100 10 g Glass bottle
AC171050500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171050100 Supplier Thermo Scientific Chemicals Supplier No. 171050100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 93-10-7 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-10-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00006752
InChI Key LOAUVZALPPNFOQ-UHFFFAOYSA-N
Synonym quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
PubChem CID 7124
ChEBI CHEBI:18386
IUPAC Name quinoline-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 159.0°C to 161.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Beilstein 22, 71
Merck Index 15, 8162
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alkali solutions,soluble in alcohol
Formula Weight 173.17
Percent Purity 99%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material Quinaldic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-218-3

RUO – Research Use Only