missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinacridone 93.0+%, TCI America™
Specifications
Specifications
| Melting Point | 390°C |
| Color | Purple-Red |
| Quantity | 25 g |
| Formula Weight | 312.33 |
| Percent Purity | ≥93.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Quinacridone |
Safety and Handling
| EINECSNumber | (5)-1167&(5)-3291 |
| RTECSNumber | VA8533500 |
| TSCA | Yes |