Learn More
Contec™ Quiltec™ I Wipes
GREENER_CHOICE
Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.
Supplier:  Contec™ QT199
 View more versions of this product
Catalog No. 19033742B
Only null left
Description
Description
Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.
  • These sealed-edge wipes offer an extremely high sorptive capacity. Two layers of quilted polyester fabric are laser-cut and sealed together to create pockets where additional fluid can be trapped.
  • Two-ply quilted polyester construction creates durable wiper with exceptional absorbency—absorbs more than twice the capacity of a single-layer wiper
  • Extremely low in both particles and fibers—ideal for interior surface cleaning of process tools, wiping down products, or general-purpose cleaning
Order Info
Laundered and packaged in an ISO Class 4 cleanroom.
Specifications
Chemical Identifiers
Cleanroom Wipe
Not Autoclavable
2-ply
Polyester
Single-use
Specifications
Cleanroom Wipe
23 cm
23 cm
Not Autoclavable
Single-use
Cleanroom
2-ply
9 in.
9 in.
Polyester
ISO Class 3 and 4
23 x 23 cm (9 x 9 in.)
50/Bag, 20 Bags/Cs.
Non-sterile
Product Suggestions
Product Suggestions