missing translation for 'onlineSavingsMsg'
Learn More

Quidel QuickVue™ RSV Test Kit

Catalog No. 23038089
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23038089 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23038089 Supplier Quidel Supplier No. Q20193

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For rapid, qualitative detection of RSV antigen directly from nasopharyngeal swab or aspirate specimens from symptomatic pediatric patients

  • Dipstick format
  • 15 Minutes to result
  • Sensitivity: 92%
  • Specificity: 92%
  • Three-step procedure requires approximately 30 seconds of hands-on time
  • Internal and external controls

Specifications

Detectable Analytes Respiratory Syncytial Virus Antigen
Format Dipstick
Sample Type Nasopharyngeal
Type RSV Test Kit
Includes All components needed to perform 20 Tests are contained in the kit, including nasopharyngeal swabs and positive and negative control
Quantity 25 Tests
Test Time 15 min.

For professional and laboratory use only