missing translation for 'onlineSavingsMsg'
Learn More

Quidel HCG Quick Urine Control for QuickVue+™ One-Step hCG Combo Lateral Flow Test Kit

Catalog No. 22043045
Change view
Click to view available options
Quantity:
2 x 4.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22043045 2 x 4.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22043045 Supplier Quidel Supplier No. Q00272
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HCG Quick Urine Control, for use with QuickVue+™ One-Step hCG Combo Lateral Flow Test Kit. 10mIU/mL sensitivity in serum; 20mIU/mL sensitivity in urine.

  • CPT Codes: 81025 (urine), 84703 (serum)
  • CLIA Complexity: Waived (urine), Non-Waived (serum)
  • Test results within five minutes (serum); three minutes (urine)
  • Two-color plus/minus (±) endpoint for easy-to-read and interpret results
  • Immunoassay for the qualitative detection of hCG in serum or urine
  • >99% sensitivity, specificity, and accuracy
  • For in vitro diagnostic use

Specifications

Accuracy >99%
Content And Storage Room Temperature
CPT Code 81025, 84703
Clia Complexity Waived, Non-waived
Detectable Analytes hCG
Sample Type Urine
Sensitivity >99%
Type HCG Quick Urine Control
Includes HCG Quick Urine Control
Quantity 2 x 4.5 mL
Specificity >99%
Test Time 3 min. for urine, 5 min. for serum
Show More Show Less