missing translation for 'onlineSavingsMsg'
Learn More

Quidel QuickVue™ In-Line™ One-Step Strep A Test

Catalog No. p-112967
Change view
Click to view available options
Quantity:
25 Tests
Type:
Streptococcus A Test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23043027 25 Tests Streptococcus A Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23043027 Supplier Quidel Supplier No. Q00343IN
Only null left

Includes

Reaction units, extraction reagents, throat swabs, package insert, and positive and negative controls

Unique patented in-line extraction method reduces hands-on time and provides quick results

  • CLIA Complexity: Waived
  • CPT Codes: 87880/87880QW
  • One-step lateral flow immunoassay for the detection of Group A streptococcal antigen from throat swab specimens
  • Results within 5 minutes
  • Sensitivity: 92%
  • Specificity: 99%
  • Overall accuracy: 96%
  • Ideal for exam room environments
  • Built-in procedural controls confirm proper test performance
  • Two-color read-out makes results easy to interpret
  • For in vitro diagnostic use

Specifications

CPT Code 87880, 87880QW
Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Sample Type Throat Swab
Type Streptococcus A Test
Includes Reaction units, Extraction reagents, Throat swabs, Package insert and Positive and Negative control
Quantity 25 Tests
Test Time 5 min.