missing translation for 'onlineSavingsMsg'
Learn More

Innovatek QuickStick™ Pregnancy Test Strips

Catalog No. 361011108
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361011108 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361011108 Supplier Innovatek Supplier No. 42204K
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovatek QuickStick™ Pregnancy Test Strips provide results within 5 minutes

  • Individual foil pouches test strip
  • Dip and read in 5 minutes

Specifications

Sample Type Urine
For Use With (Application) Pregnancy Testing
Packaging Type Individually Packaged
Quantity 50 Tests
Test Time ≈5 min.