missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Innovatek QuickStep™ Plus Pregnancy Cassette Tests
Description
- Patient ID can be labeled on each cassette
- For urine, or urine/serum testing
- Individually foil wrapped, includes pipette and desiccant
- Pipette sample into well and read in 5 minutes
Specifications
Specifications
| Sample Type | Urine/Serum |
| For Use With (Application) | Pregnancy Testing |
| Quantity | 20 Tests |
| Test Time | ≈5 min. |