Learn More
Hardy Diagnostics™ QuickSlide™ Replacement Tubing Kit for HemaPRO Hematology Stainer
Description
- A tubing change is recommended at least once annually.
- The kit consists of labeled tubing, (4) orange pump tubes, reagent cannulas, check valves, tubing elbows and the wire ties to secure the tubing to the unit.
- Tubing can be installed in just a few minutes
Specifications
Specifications
| Product Type | Replacement Tubing Kit |
| Description | Weight: 1lb., L X W X H: 12 X 7 X 3 in. |
| For Use With (Equipment) | For use with QS-001, and QS-29000 |