missing translation for 'onlineSavingsMsg'
Learn More

Contec QUICKCONNECT PRESS FIT ADAPTOR

Catalog No. 17103015
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
17103015 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17103015 Supplier Contec Supplier No. CTC6084

The QuickConnect™ Adaptor allows users to connect any QuickConnect handle to the Window Cleaning Squeegee.

  • For use with QuickConnect handles and Window Cleaning Squeegee
  • Autoclavable at 121°C for 30 minutes

Specifications

Product Type Handle, Mop Adapter
Color Code Black
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Color-coded No
Material Stainless Steel