missing translation for 'onlineSavingsMsg'
Learn More

Contec™ QuickConnect™ Mop Head Frames
GREENER_CHOICE

Catalog No. 19176941
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
VertiKlean MAX Mop Wipe System
Length (Metric):
29 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Length (Metric)
19176941 VertiKlean MAX Mop Wipe System 29 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 19176941 missing translation for 'mfr' Contec™ missing translation for 'supplierNo' 2652K

Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ QuickConnect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.

  • Offer maximum flexibility for cleaning in critical environments.
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable

Specifications

Color Code White
Product Type Mop Frame
Length (English) 11.5 in.
Sterility Non-sterile
Length (Metric) 29 cm
Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) VertiKlean MAX Mop Wipe System
For Use With (Application) Pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments
Color-coded No
Material Stainless Steel
Show More Show Less