Learn More
Innovatek QuickCheck Urine Drug Screen Tests
Description
- Accurate and sensitive test procedure
- Built-in test controls
- Clear, easy-to-read, precise test results within 5 minutes
- Meets US SAMSHA and DOT requirements
- Creatinine test (CRE) included
- Temperature indicator on drug screen cups
Specifications
Specifications
| Certifications/Compliance | US SAMSHA, DOT |
| Detectable Analytes | Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Creatinine, Methodone (MTD), Opiates (OPI), Oxycodone (OXY), Cannabinoids (THC), Methamphetamine (mAMP), Phencyclidine (PCP) |
| Type | 12 Cup |
| Disposable | Single-use |
| For Use With (Application) | Drug Testing |
| Test Time | 5 min. |