missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuickBag™ Buffered Peptone Water

Catalog No. p-8015464
Change view
Click to view available options
Quantity:
2 x 4.5 L
3 x 2.7 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DQ0509A 3 x 2.7 L
DQ0509B 2 x 4.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DQ0509A Supplier Thermo Scientific™ Supplier No. DQ0509A
Only null left

Shorten your workflow with easy to prepare 2.7L or 4.5L media bags: simply merge the segments to reconstitute in two minutes.

Save time in preparing selective enrichment medium for the isolation of Salmonella spp. from food samples using Thermo Scientific™ QuickBag™ Buffered Peptone Water. The irradiated medium is supplied in transparent plastic bags. Simply pop the seal to mix the water, and medium segments to reconstitute in two minutes. QuickBag Enrichment Media are available in 2.7L and 4.5L sizes to suit your workflow. Medium can be dispensed through the outlet tube as required.

Shorten your workflow with FitBag UVM1 Broth. FitBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

  • Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

Specifications

Content And Storage 10°C to 30°C
Detects QuickBag Buffered Peptone Water
Format Convenience Format
Product Type Pre-enrichment Medium
Quantity 3 x 2.7 L