Learn More
Chemglass Life Sciences Column, Chromatography, Quick Separation Funnel, 90mm Coarse Frit, 305mm height above the disc, 24/40 Lower Vacuum Assembly
Supplier: Chemglass Inc CG141236
Column, Chromatography, Quick Separation Funnel, 90mm Coarse Frit, 305mm height above the disc, 24/40 Lower Vacuum Assembly. For performing quick separations using a vacuum source. All funnels have a lower standard taper inner joint with a sealed in drip tip and an outlet for connection to a vacuum source. All sizes have a Coarse porosity fritted disc. Special sizes are available, please call for details.