missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Quick Fluorescent Loading Buffer for SDS-PAGE, 3X

Catalog No. ICN08W00013
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN08W00013 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN08W00013 Supplier MP Biomedicals™ Supplier No. 08W00013

Quick Fluorescent Loading Buffer for SDS-PAGE comes in a reducing format with DTT and is more sensitive than silver staining.

The Quick Fluorescent Loading Buffer pre-stains a protein sample for SDS-PAGE. Protein samples are stained fluorescently during sample treatment prior to electrophoresis. Protein bands in an SDS-gel or on a membrane after transfer can be visualized and analyzed by a UV or LED transilluminator or a digital gel imaging system.

  • Fluorescently pre-stains protein samples prior to gel electrophoresis
  • Visualize proteins directly within the gel using a UV or LED transilluminator

Specifications

Buffer Loading Buffer
Color Clear
Concentration 3X
Detection Method Electrophoresis
Gel Type SDS-PAGE
Volume (Metric) 1 mL
Form Liquid
Quantity 1 Vial
Storage Requirements -20°C
Product Type Loading Buffer

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.