Learn More
Research Products Quick-DNA MiniPrep w/Zymo-Spin IIC Columns, Capped, 200 Preps
Supplier: Research Products Corporation ZD3025
The Quick-DNA Kits are ideal DNA isolation kits for easy, rapid isolation of total DNA (e.g., genomic, mitochondrial, viral) from a variety of biological sample sources. Whole blood (fresh or stored), buffy coat, buccal cells, cells from culture, saliva, and other biological liquid samples can be processed with these kits. Zymo-Spin Column/Plate technology enables high-quality DNA purification in minutes. PCR inhibitors are effectively removed, and the eluted DNA is ideal for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.
Easy purification of high-quality DNA from whole blood, buffy coat, swabs, or cultured cells.
Protocol excludes the use of Proteinase K and organic denaturants for biofluid and cell samples.
Eluted, inhibitor-free DNA is ideal for PCR, endonuclease digestion, bisulfite conversion/methylation detection, sequencing, genotyping, etc.