Learn More
New England Biolab Ltd Quick CIP 5000 units DFS Item
Supplier:  New England Biolab Ltd M0525L
Quick CIP is a heat-labile recombinant version of calf intestinal alkaline phosphatase (CIP) purified from a recombinant source. Quick CIP nonspecifically catalyzes the dephosphorylation of 5 and 3 ends of DNA and RNA phosphomonoesters. It also hydrolyses ribo- and deoxyribonucleoside triphosphates (NTPs and dNTPs). Quick CIP is useful in many applications that require the dephosphorylation of DNA or RNA ends. In cloning, dephosphorylation prevents re-ligation of linearized plasmid DNA. The enzyme can quickly dephosphorylate 5 protruding, 5 recessed, and blunt ends in just 10 minutes. Quick CIP may also be used to degrade unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis.
Numéro de catalogue. NC1738046
Il en reste null
Découvrez plus d'offres spéciales