missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine Hemifumarate 98.0+%, TCI America™

Catalog No. p-7135648
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00925G 5 g
Q009225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00925G Supplier TCI America Supplier No. Q00925G
Only null left

Chemical Identifiers

CAS 111974-72-2
Molecular Formula C29H33N3O10S
Molecular Weight (g/mol) 615.65
MDL Number MFCD03423782
InChI Key JLWSQVHJLLYDPX-LVEZLNDCSA-N
Synonym quetiapine fumarate, 2-methoxyethanol; quetiapine
PubChem CID 45358180
IUPAC Name bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
SMILES OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 5 g
Formula Weight 441.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Quetiapine Hemifumarate
RTECSNumber KK3605000
TSCA No