missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quercetin dihydrate, 99.85%, For HPLC analysis, MP Biomedicals™
$104.42 - $104.42
Chemical Identifiers
| CAS | 6151-25-3 |
|---|---|
| Molecular Formula | C15H14O9 |
| Molecular Weight (g/mol) | 338.27 |
| MDL Number | MFCD00149487 |
| InChI Key | GMGIWEZSKCNYSW-UHFFFAOYSA-N |
| Synonym | quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate |
| PubChem CID | 5284452 |
| IUPAC Name | 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate |
| SMILES | O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15200325
|
MP Biomedicals Inc
0215200325 |
25 g |
Each for $104.42
|
|
|||||
Chemical Identifiers
| 6151-25-3 | |
| 338.27 | |
| GMGIWEZSKCNYSW-UHFFFAOYSA-N | |
| 5284452 | |
| O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1 |
| C15H14O9 | |
| MFCD00149487 | |
| quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate | |
| 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate |
Specifications
| 6151-25-3 | |
| C15H14O9 | |
| 25 g | |
| GMGIWEZSKCNYSW-UHFFFAOYSA-N | |
| 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate | |
| 5284452 | |
| 0.9985 | |
| Quercetin dihydrate, Molecular Biology |
| Green-Yellow | |
| MFCD00149487 | |
| quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate | |
| O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1 | |
| 338.27 | |
| 338.26 | |
| Powder |
Safety and Handling
EINECSNumber : 204-187-1
Recommended Storage : Store at +4°C.