missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quercetin dihydrate, 99.85%, For HPLC analysis, MP Biomedicals™
Specifications
Specifications
| Color | Green-Yellow |
| Quantity | 25 g |
| Formula Weight | 338.26 |
| Percent Purity | 0.9985 |
| Physical Form | Powder |
| Chemical Name or Material | Quercetin dihydrate, Molecular Biology |
Safety and Handling
| EINECSNumber | 204-187-1 |
| Recommended Storage | Store at +4°C. |